ENA238710481 Enamine

[3-(dimethylamino)-2-{[(dimethylamino)methylidene]amino}prop-2-en-1-ylidene]dimethylazanium perchlorate

  • Cas Number : 50704-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Purity 95
Salt Data ClO4-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35813
Availability details for
Quantity
  • Availability
  • Add to Cart