ENA238710482 Enamine

N'-(2-cyanophenyl)-N,N-dimethylmethanimidamide

  • MDL number MFCD00456540
  • Cas Number : 1227920-69-5
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36115
Availability details for
Quantity
  • Availability
  • Add to Cart