ENA238710497 Enamine

N1-ethylbenzene-1,2-diamine

  • MDL number MFCD11186962
  • Cas Number : 23838-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37435
Availability details for
Quantity
  • Availability
  • Add to Cart