ENA238710501 Enamine

2-[(diethylamino)methyl]aniline

  • MDL number MFCD08144100
  • Cas Number : 27958-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39085
Availability details for
Quantity
  • Availability
  • Add to Cart