ENA238710514 Enamine

2,6-dichloro-4-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD09738804
  • Cas Number : 74875-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40129
Availability details for
Quantity
  • Availability
  • Add to Cart