ENA238710539 Enamine

2-fluoro-3-nitropyridine

  • MDL number MFCD03095068
  • Cas Number : 1480-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41103

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-41103
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart