ENA238710542 Enamine

2-(3-methanesulfonylphenoxy)acetic acid

  • MDL number MFCD12065103
  • Cas Number : 1878-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-113
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41303
Availability details for
Quantity
  • Availability
  • Add to Cart