ENA238710574 Enamine

2-{[(pyridin-3-yl)methyl]amino}propanoic acid dihydrochloride

  • MDL number MFCD11857884
  • Cas Number : 1396962-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42240
Availability details for
Quantity
  • Availability
  • Add to Cart