ENA238710583 Enamine

2-(morpholin-4-yl)-6H,7H-pyrimido[4,5-d][1,3]thiazol-7-one

  • MDL number MFCD09041564
  • Cas Number : 99967-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 397...399
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42433
Availability details for
Quantity
  • Availability
  • Add to Cart