ENA238710598 Enamine

5-(4-butoxyphenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182393
  • Cas Number : 334504-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42661
Availability details for
Quantity
  • Availability
  • Add to Cart