ENA238710600 Enamine

pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD00040385
  • Cas Number : 6345-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234...236
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42664

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42664
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart