ENA238710603 Enamine

pyridine-3-carboximidamide hydrochloride

  • MDL number MFCD00054326
  • Cas Number : 7356-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42667

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42667
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart