ENA238710605 Enamine

1-[(4-aminophenyl)methyl]pyrrolidin-2-one

  • MDL number MFCD06740236
  • Cas Number : 245546-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42676

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42676
Availability details for
Quantity
  • Availability
  • Add to Cart