ENA238710609 Enamine

2-[3-(cyclohex-1-en-1-yl)-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl]acetic acid

  • MDL number MFCD11857942
  • Cas Number : 1146289-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42687
Availability details for
Quantity
  • Availability
  • Add to Cart