ENA238710612 Enamine

2-(4-chlorophenyl)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD08442919
  • Cas Number : 926187-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42706

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42706
Availability details for
Quantity
  • Availability
  • Add to Cart