ENA238710613 Enamine

N-[2-(dimethylamino)ethyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxamide dihydrochloride

  • MDL number MFCD11857943
  • Cas Number : 1214014-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42707
Availability details for
Quantity
  • Availability
  • Add to Cart