ENA238710616 Enamine

5-(naphthalen-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD08753498
  • Cas Number : 198780-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42719
Availability details for
Quantity
  • Availability
  • Add to Cart