ENA238710620 Enamine

tert-butyl 4-[(2,2,2-trifluoroethyl)amino]piperidine-1-carboxylate

  • MDL number MFCD11857945
  • Cas Number : 1146289-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42731

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42731
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart