ENA238710627 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[3,4-a]pyridin-3-ylmethanamine

  • MDL number MFCD08691462
  • Cas Number : 915923-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42738

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-42738
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart