ENA238710628 Enamine

6-(1H-1,2,4-triazol-1-yl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD11857950
  • Cas Number : 1146289-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42741
Availability details for
Quantity
  • Availability
  • Add to Cart