ENA238710641 Enamine

3-(3,4-dichlorophenyl)-2-oxopropanoic acid

  • MDL number MFCD09729220
  • Cas Number : 128437-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42761
Availability details for
Quantity
  • Availability
  • Add to Cart