ENA238710644 Enamine

4-(3-bromothiophen-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD00793890
  • Cas Number : 81216-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42765
Availability details for
Quantity
  • Availability
  • Add to Cart