ENA238710645 Enamine

2-(2-phenyl-2H-1,2,3-triazol-4-yl)acetic acid

  • MDL number MFCD11857954
  • Cas Number : 64404-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42766

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42766
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart