ENA238710670 Enamine

1-(4-chloro-1H-pyrrole-2-carbonyl)piperazine

  • MDL number MFCD11167893
  • Cas Number : 1042557-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42811
Availability details for
Quantity
  • Availability
  • Add to Cart