ENA238710679 Enamine

4-(2-bromophenoxy)butanenitrile

  • MDL number MFCD09932003
  • Cas Number : 1016513-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42823
Availability details for
Quantity
  • Availability
  • Add to Cart