ENA238710683 Enamine

1-(cyclohexylcarbamoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD04212920
  • Cas Number : 674299-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42831

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42831
Availability details for
Quantity
  • Availability
  • Add to Cart