ENA238710685 Enamine

3,7-diethyl 6-(3-hydroxypropyl)pyrazolo[1,5-a]pyrimidine-3,7-dicarboxylate

  • MDL number MFCD11857966
  • Cas Number : 1146289-94-2
  • Molecular Weight : 321.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42833
Availability details for
Quantity
  • Availability
  • Add to Cart