ENA238710695 Enamine

5,7-dichloro-3-hydrazinylidene-2,3-dihydro-1H-indol-2-one

  • Cas Number : 35246-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03689
Availability details for
Quantity
  • Availability
  • Add to Cart