ENA238710698 Enamine

4-(chloromethyl)-1-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD11857973
  • Cas Number : 177550-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42863
Availability details for
Quantity
  • Availability
  • Add to Cart