ENA238710713 Enamine

3-(heptylamino)propan-1-ol

  • MDL number MFCD06408487
  • Cas Number : 1024275-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42883
Availability details for
Quantity
  • Availability
  • Add to Cart