ENA238710736 Enamine

[3-(2-methoxyphenyl)-1,2,4-oxadiazol-5-yl]methanol

  • MDL number MFCD08442623
  • Cas Number : 926242-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42927
Availability details for
Quantity
  • Availability
  • Add to Cart