ENA238710738 Enamine

3-(pyrrolidin-1-yl)piperidine

  • MDL number MFCD09998985
  • Cas Number : 144243-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42929
Availability details for
Quantity
  • Availability
  • Add to Cart