ENA238710745 Enamine

2-[3-(1-aminoethyl)phenoxy]-N-ethylacetamide

  • MDL number MFCD09736561
  • Cas Number : 954279-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42948
Availability details for
Quantity
  • Availability
  • Add to Cart