ENA238710755 Enamine

2-(4-bromophenyl)-1$l^{6},2-thiazolidine-1,1-dione

  • MDL number MFCD09933347
  • Cas Number : 71703-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42958

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42958
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart