ENA238710758 Enamine

7-(trifluoromethyl)-1,4-diazepan-5-one

  • MDL number MFCD11858217
  • Cas Number : 1146290-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42963
Availability details for
Quantity
  • Availability
  • Add to Cart