ENA238710764 Enamine

3-phenyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD00003159
  • Cas Number : 17467-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42973

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-42973
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart