ENA238710765 Enamine

2-[4-methyl-6-oxo-2-(pyridin-4-yl)-1,6-dihydropyrimidin-5-yl]acetic acid

  • MDL number MFCD11857997
  • Cas Number : 1146290-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42974
Availability details for
Quantity
  • Availability
  • Add to Cart