ENA238710774 Enamine

2-amino-2-(2,4-dichlorophenyl)acetonitrile

  • MDL number MFCD09729814
  • Cas Number : 189138-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42983
Availability details for
Quantity
  • Availability
  • Add to Cart