ENA238710776 Enamine

N-(3-amino-4-chlorophenyl)propanamide

  • MDL number MFCD04104705
  • Cas Number : 213831-01-7
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42986
Availability details for
Quantity
  • Availability
  • Add to Cart