ENA238710777 Enamine

3-(thiophene-2-sulfonamido)benzoic acid

  • MDL number MFCD00741837
  • Cas Number : 82068-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42987
Availability details for
Quantity
  • Availability
  • Add to Cart