ENA238710783 Enamine

2,3-dichloroquinoxaline-6-sulfonyl chloride

  • MDL number MFCD11858002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42995
Availability details for
Quantity
  • Availability
  • Add to Cart