ENA238710784 Enamine

2-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)acetic acid

  • MDL number MFCD08752443
  • Cas Number : 104189-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-240
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42996
Availability details for
Quantity
  • Availability
  • Add to Cart