ENA238710785 Enamine

2-(5-bromothiophen-2-yl)acetonitrile

  • MDL number MFCD01859859
  • Cas Number : 71637-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43001
Availability details for
Quantity
  • Availability
  • Add to Cart