ENA238710789 Enamine

ethyl 2-[2-(methylimino)-3-(2,4,6-trimethylphenyl)-2,3-dihydro-1,3-thiazol-4-yl]acetate

  • MDL number MFCD06654940
  • Cas Number : 885459-15-4
  • Molecular Weight : 318.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06424
Availability details for
Quantity
  • Availability
  • Add to Cart