ENA238710795 Enamine

2-amino-N-(5-chloropyridin-2-yl)benzamide

  • MDL number MFCD11184859
  • Cas Number : 280771-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43025
Availability details for
Quantity
  • Availability
  • Add to Cart