ENA238710798 Enamine

4-amino-N-ethylbenzamide

  • MDL number MFCD01569372
  • Cas Number : 89399-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43029
Availability details for
Quantity
  • Availability
  • Add to Cart