ENA238710815 Enamine

2,4-dichloro-N-[(pyridin-3-yl)methyl]aniline

  • MDL number MFCD00113737
  • Cas Number : 84324-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43054
Availability details for
Quantity
  • Availability
  • Add to Cart