ENA238710819 Enamine

6-hydroxy-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD08689914
  • Cas Number : 116569-08-5
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43058
Availability details for
Quantity
  • Availability
  • Add to Cart