ENA238710823 Enamine

tert-butyl N-(4-amino-2-methoxyphenyl)carbamate

  • MDL number MFCD09808305
  • Cas Number : 190851-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43064
Availability details for
Quantity
  • Availability
  • Add to Cart