ENA238710831 Enamine

N-(2-butyl-1H-1,3-benzodiazol-5-yl)acetamide

  • MDL number MFCD11858014
  • Cas Number : 1146290-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192...194
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43129
Availability details for
Quantity
  • Availability
  • Add to Cart